Learn More
Selleck Chemical LLC Desipramine Hydrochloride
SDP
Catalog No. 5000003464
Encompass_Preferred
Desipramine hydrochloride (Desmethylimipramine Norimipramine EX-4355 G-35020 JB-8181 NSC-114901) is a dibenzazepine-derivative tricyclic antidepressant that acts as a selective norepinephrine reuptake inhibitor It also shows weak serotonin reuptake inhibitory 1-blocking antihistamine and anticholinergic effects
Catalog No. 50-000-03464 Supplier Selleck Chemical LLC Supplier No. S5485100MG
Desipramine hydrochloride (Desmethylimipramine Norimipramine EX-4355 G-35020 JB-8181 NSC-114901) is a dibenzazepine-derivative tricyclic antidepressant that acts as a selective norepinephrine reuptake inhibitor It also shows weak serotonin reuptake inhibitory 1-blocking antihistamine and anticholinergic effects