Learn More
Medchemexpress LLC HY-111490 10mg Medchemexpress, Desisobutyryl-ciclesonide CAS:161115-59-9 Purity:>98%
SDP
Catalog No. 501968884
Encompass_Preferred
Medchemexpress, HY-111490 10mg Desisobutyryl-ciclesonide CAS:161115-59-9 Desisobutyryl-ciclesonide is the active metabolite of Ciclesonide. Desisobutyryl-ciclesonide has affinity for the glucocorticoid receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8884 Supplier Medchemexpress LLC Supplier No. HY11149010MG
Medchemexpress, HY-111490 10mg Desisobutyryl-ciclesonide CAS:161115-59-9 Desisobutyryl-ciclesonide is the active metabolite of Ciclesonide. Desisobutyryl-ciclesonide has affinity for the glucocorticoid receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.