Learn More
Cole-Parmer Traceable Single-Display Triple-Event Lab Digital Timer with Calibration
Catalog No. NC0073741 Shop All Cole-Parmer Products
Encompass_Preferred
Each timing channel has a distinct alarm and display indicator ideal for drying and incubating operations. Alarm can be shut off manually, or shuts off automatically after one minute. Compact, lightweight timer is made of chemical-resistant ABS plastic. Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Catalog No. NC0073741 Supplier Cole-Parmer Supplier No. 9446060
Each timing channel has a distinct alarm and display indicator ideal for drying and incubating operations. Alarm can be shut off manually, or shuts off automatically after one minute. Compact, lightweight timer is made of chemical-resistant ABS plastic. Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.