Learn More
Bevco Precision Manufacturing Co Versa Desk Height Polyurethane Backless Stool; Black Nylon Base
Catalog No. NC2022935 Shop All Bevco Precision Manufacturing Co Products
Encompass_Preferred
BEVCO's sturdy construction and modern design of the Versa backless stool is ideal for a wide range of applications, including general office, medical, laboratory and factory. The 14" diameter, 2" thick black polyurethane seat is impervious to water, oil, and most chemicals. The non-slip surface is puncture resistant and easy to clean. The pneumatic height adjustment allows for stress-free seat height changes from 15" - 20". The stool features a sturdy 25" diameter black nylon base. Model comes standard with mushroom glides. Optional casters can be ordered separately.
Catalog No. NC2022935 Supplier Bevco Precision Manufacturing Co Supplier No. 3000P
BEVCO's sturdy construction and modern design of the Versa backless stool is ideal for a wide range of applications, including general office, medical, laboratory and factory. The 14" diameter, 2" thick black polyurethane seat is impervious to water, oil, and most chemicals. The non-slip surface is puncture resistant and easy to clean. The pneumatic height adjustment allows for stress-free seat height changes from 15" - 20". The stool features a sturdy 25" diameter black nylon base. Model comes standard with mushroom glides. Optional casters can be ordered separately.