Learn More
Electron Microscopy Sciences One Counting Unit Tally Counter
SDP
Catalog No. 5025325
Encompass_Preferred
Chrome plated lab counter with counter with a flange (2 1/2" dia) for desktop mounting. Recording lever is on the right side and the reset knob on the top. The counter is ideal for serial sectioning work.
Catalog No. 50-253-25 Supplier Electron Microscopy Sciences Supplier No. 70310
Chrome plated lab counter with counter with a flange (2 1/2" dia) for desktop mounting. Recording lever is on the right side and the reset knob on the top. The counter is ideal for serial sectioning work.