Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Desktop Beta Box
SCI_ED

Catalog No. S29143 Shop All Fisherbrand Products
Change view
Click to view available options
Height (Metric):
15.2 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
S29143 15.2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S29143

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For holding tubes and racks

  • Made from 0.37 in. thick (0.95 cm) acrylic
  • Cover has handle
  • Base has four nonskid feet
  • Dimensions: 6L x 6W x 6 in.H (15.2 x 15.2 x 15.2 cm)

Specifications

Material Acrylic
Dimensions (L x W x H) 6 x 6 x 6 in. (15.2 x 15.2 x 15.2 cm)
For Use With (Application) To store various racks, reagent bottles, and other vessels containing beta-emitting isotopes
Height (English) 6 in.
Height (Metric) 15.2 cm
Holds Tubes and racks
Length (English) 6 in.
Includes Lid and radiation warning labels, 4 non-skid feet
Length (Metric) 15.2 cm
Product Type Desktop Beta box
Thickness (English) 0.37 in.
Width (English) 6 in.
Width (Metric) 15.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.