Learn More
RWD LIFE SCIENCE INC Digital Display Stereotaxic Instrument
SDP
Catalog No. NC3388876
Encompass_Preferred
RWD desktop digital stereotaxic instrument adds a digital display module on the basis of the standard type. The displacement sensor can display the displacement more accurately, reducing the tedious steps of manual reading. The displacement accuracy reaches 10 um, and the origin can be set arbitrarily within the range.
Catalog No. NC3388876 Supplier RWD LIFE SCIENCE INC Supplier No. 68803
Add to Cart
Add to Cart
Request a Quote
RWD desktop digital stereotaxic instrument adds a digital display module on the basis of the standard type. The displacement sensor can display the displacement more accurately, reducing the tedious steps of manual reading. The displacement accuracy reaches 10 um, and the origin can be set arbitrarily within the range.