Learn More
Description
- Available in polymer or wood or metal options, featuring a transparent design for easy card inspection without removal.
- Compatible with A4 cards
Specifications
Specifications
| Product Type | Fingerprint Cardholder |
| For Use With (Application) | Fingerprinting |
| Format | Triple |
| Material | Wood, Metal |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
