Learn More
Cayman Chemical Deslanoside, Purity: ≥98%, CAS: 17598-65-1, 5 mg
SDP
Catalog No. 502989259
Encompass_Preferred
A cardiac glycoside; induces ventricular tachycardia in anesthetized dogs at 0.18 mg/kg
Catalog No. 50-298-9259 Supplier Cayman Chemical Supplier No. 308725 MG
A cardiac glycoside; induces ventricular tachycardia in anesthetized dogs at 0.18 mg/kg