Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deslanoside (desacetyllanatoside C) | 17598-65-1 | MFCD00135818 | 99.8% | 943.08 g/mol | C47H74O19 | 10 MG
SDP

Catalog No. 5000299287
Encompass_Preferred

Deslanoside is a cardiac glycoside (desacetyllanatoside C) used as a research reagent for studies of cardiac function, ion transport, and Na+/K+-ATPase inhibition. It is supplied as a high-purity solid and is also available as a DMSO solution for biochemical assays.

  • Cardiac glycoside for research use.
  • High purity (99.79%).
  • Molecular weight 943.08 g/mol.
  • Chemical formula C47H74O19.
  • CAS number 17598-65-1.
  • Available as 10 MG solid and 10 mM in DMSO solution.
  • Suitable for biochemical and pharmacological studies of arrhythmia mechanisms and ion transport.

Catalog No. 50-002-99287 Supplier Medchemexpress LLC Supplier No. HYA015410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deslanoside is a cardiac glycoside (desacetyllanatoside C) used as a research reagent for studies of cardiac function, ion transport, and Na+/K+-ATPase inhibition. It is supplied as a high-purity solid and is also available as a DMSO solution for biochemical assays.

  • Cardiac glycoside for research use.
  • High purity (99.79%).
  • Molecular weight 943.08 g/mol.
  • Chemical formula C47H74O19.
  • CAS number 17598-65-1.
  • Available as 10 MG solid and 10 mM in DMSO solution.
  • Suitable for biochemical and pharmacological studies of arrhythmia mechanisms and ion transport.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.