Learn More
Cayman Chemical Deslanoside, Purity: ≥98%, CAS: 17598-65-1, 10 mg
SDP
Catalog No. 502990024
Encompass_Preferred
A cardiac glycoside; induces ventricular tachycardia in anesthetized dogs at 0.18 mg/kg
Catalog No. 50-299-0024 Supplier Cayman Chemical Supplier No. 3087210 MG
A cardiac glycoside; induces ventricular tachycardia in anesthetized dogs at 0.18 mg/kg