Learn More
Selleck Chemical LLC Desloratadine 10mg 100643-71-8 SCH-34117, NSC 675447
SDP
Catalog No. 501363534
Encompass_Preferred
Desloratadine (SCH-34117, NSC 675447) is a potent antagonist for human histamine H1 receptor with IC50 of 51 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3534 Supplier Selleck Chemical LLC Supplier No. S401210MG
Desloratadine (SCH-34117, NSC 675447) is a potent antagonist for human histamine H1 receptor with IC50 of 51 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.