Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Desloratadine

Catalog No. AC462920010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462920010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC462920010 Supplier Thermo Scientific Chemicals Supplier No. 462920010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100643-71-8 | C19H19ClN2 | 310.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100643-71-8
Molecular Formula C19H19ClN2
Molecular Weight (g/mol) 310.83
InChI Key JAUOIFJMECXRGI-UHFFFAOYSA-N
IUPAC Name 13-chloro-2-(piperidin-4-ylidene)-4-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene
SMILES ClC1=CC=C2C(CCC3=CC=CN=C3C2=C2CCNCC2)=C1

Specifications

Melting Point 150.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: slightly soluble. Other solubilities: soluble in ethanol, propylene glycol, dmso
Formula Weight 310.82
Chemical Name or Material Desloratadine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.