Learn More
Medchemexpress LLC Desloratadine-d5 | 1020719-34-9 | >99.7% | 1 MG
SDP
Catalog No. 5000405488
Encompass_Preferred
Desloratadine-d5 is a deuterium-labeled version of Desloratadine, an orally active major metabolite of the nonsedating H1-antihistamine Loratadine. It functions as a selective H1-receptor antagonist with anti-allergic and anti-inflammatory activities.
  • Used as a tracer for quantitative analysis
  • Acts as a selective H1-receptor antagonist
  • Possesses anti-allergic and anti-inflammatory properties
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for drug development and quantitation
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-05488 Supplier Medchemexpress LLC Supplier No. HYB0539S31MG
Desloratadine-d5 is a deuterium-labeled version of Desloratadine, an orally active major metabolite of the nonsedating H1-antihistamine Loratadine. It functions as a selective H1-receptor antagonist with anti-allergic and anti-inflammatory activities.
  • Used as a tracer for quantitative analysis
  • Acts as a selective H1-receptor antagonist
  • Possesses anti-allergic and anti-inflammatory properties
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for drug development and quantitation
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs