Learn More
Cayman Chemical Desloratadine-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2713301-38-1, 1 mg
SDP
Catalog No. 502935209
Encompass_Preferred
An internal standard for the quantification of desloratadine by GC- or LC-MS
Catalog No. 50-293-5209 Supplier Cayman Chemical Supplier No. 267761 MG
An internal standard for the quantification of desloratadine by GC- or LC-MS