Learn More
CPC Scientific Pyr-His-Trp-Ser-Tyr-D-Trp-Leu-Arg-Pro-NHEt
(acetate salt) 25MG
SDP
Supplier:  CPC Scientific LHRH016B
Encompass
Short-term administration of deslorelin stimulates the formation of LH and FSH, which induce an increase in the production of testosterone and estradiol. Prolonged administration of the GnRH agonist causes a sustained down-regulation of circulating LH and FSH levels and hence suppression of steroidogenesis.ONE-LETTER SEQUENCE: Pyr-HWSYwLRP-NHEtMOLECULAR FORMULA: C64H83N17O12MOLECULAR WEIGHT:1282.47STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [57773-65-6], RESEARCH AREA: HormonalREFERENCES: D.H.Coy et al., J. Med. Chem., 19, 423 (1976); K.Sundaram et al., Life Sci., 28, 83 (1981); B.Candas et al., J. Clin. Endocrinol. Metab., 70, 1046 (1990); P.P.Feuillan et al., J. Endocrinol. Invest., 24, 734 (2001); Merck Index, 14th ed., No. 2923, (2006)
Catalog No. 50-210-9668