Learn More
ILEX LIFE SCIENCES LLC Deslorelin (GnRH agonist), Synthetic Peptide, 25 mg
SDP
Supplier:  ILEX LIFE SCIENCES LLC A2240025
Encompass_Preferred
Deslorelin is a synthetic nonapeptide analogue of the natural gonadotropin releasing hormone (GnRH) with potential antineoplastic activity. Deslorelin binds to and activates pituitary gonadotropin releasing hormone (GnRH) receptors. Deslorelin has a molecular weight of 1281.64 Dalton. MOLECULAR FORMULA: C64H83N17O12. AMINO ACID SEQUENCE: pGlu-His-Trp-Ser-Tyr-D-Trp-Leu-Arg-Pro-NHC2H5. CAS REGISTRY NO.: 57773-65-6. PURITY: Greater than 99.0%. FORMULATION: Sterile-filtered white lyophilized (freeze-dried) powder. The Deslorelin peptide was lyophilized with no additives. OTHER NAMES: Gonadotropin-releasing hormone (GnRH) superagonist, luteinizing hormone-releasing hormone agonist, luteinising hormone-releasing hormone agonist, GnRH agonist, LHRH agonist.
Catalog No. 50-233-9580