Learn More
World Precision Instrument Desmarres Lid Retractor, 14 cm, 15 mm wide Blade
SDP
Catalog No. 502538344
Encompass
DeMarres Lid Retractors are surgical instruments used in ophthalmic surgery to hold back the eyelids and expose the eye for examination or surgical procedures. The DeMarres lid retractor is designed to provide a clear and unobstructed view of the eye, particularly during surgeries involving the anterior segment of the eye. The blade of the rectractor is curved to follow the contour of the eye, giving optimal exposure with limited discomfort. The tip of the blade is rounded and blunt to avoid injury to delicate eye tissue. Choose the size of the retractor you need: 0 (11 mm wide), 1 (13 mm wide), and 2 (15 mm wide). The square handles are lightly textured and polished for a solid grip. These retractors are made of surgical grade stainless steel and are suitable for autoclaving.
Catalog No. 50-253-8344 Supplier World Precision Instrument Supplier No. WP108230
DeMarres Lid Retractors are surgical instruments used in ophthalmic surgery to hold back the eyelids and expose the eye for examination or surgical procedures. The DeMarres lid retractor is designed to provide a clear and unobstructed view of the eye, particularly during surgeries involving the anterior segment of the eye. The blade of the rectractor is curved to follow the contour of the eye, giving optimal exposure with limited discomfort. The tip of the blade is rounded and blunt to avoid injury to delicate eye tissue. Choose the size of the retractor you need: 0 (11 mm wide), 1 (13 mm wide), and 2 (15 mm wide). The square handles are lightly textured and polished for a solid grip. These retractors are made of surgical grade stainless steel and are suitable for autoclaving.