Learn More
Cayman Chemical Desmethyl Bosentan, Purity: ≥95%, CAS: 253688-61-8, 5 mg
SDP
Catalog No. 502946752
Encompass_Preferred
An active metabolite of bosentan; activates the PXR in a reporter assay at 25 µM
Catalog No. 50-294-6752 Supplier Cayman Chemical Supplier No. 341575 MG
An active metabolite of bosentan; activates the PXR in a reporter assay at 25 µM