Learn More
Cayman Chemical Desmethyl Carvedilol, Purity: ≥90%, CAS: 72956-44-6, 10 mg
SDP
Catalog No. 502909582
Encompass_Preferred
An active metabolite of carvedilol; inhibits store-overload-induced calcium release in HEK293 cells expressing RyR2R4496C (IC50 = 7.62 µM); reduces increases in heart rate and prevents decreases in diastolic blood pressure induced by isoproterenol in conscious rabbits (ED50s = 32 and 5 µg/kg)
Catalog No. 50-290-9582 Supplier Cayman Chemical Supplier No. 3541110 MG
An active metabolite of carvedilol; inhibits store-overload-induced calcium release in HEK293 cells expressing RyR2R4496C (IC50 = 7.62 µM); reduces increases in heart rate and prevents decreases in diastolic blood pressure induced by isoproterenol in conscious rabbits (ED50s = 32 and 5 µg/kg)