Learn More
Medchemexpress LLC HY-13256A 5mg Medchemexpress, Desmethyl Erlotinib CAS:183321-86-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13256A5MG
Encompass_Preferred
Medchemexpress, HY-13256A 5mg Desmethyl Erlotinib CAS:183321-86-0 Desmethyl Erlotinib (OSI-420 free base) is an active metabolite of Erlotinib. Erlotinib is a potent EGFR tyrosin kinase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1317