Learn More
Medchemexpress LLC Desmethyl erlotinib (OSI-420 free base) | 183321-86-0 | MFCD09841129 | 98.8% | C21H21N3O4 | 10MG
SDP
Catalog No. 5000261049
Encompass_Preferred
Desmethyl erlotinib is the primary active metabolite of the epidermal growth factor receptor (EGFR) inhibitor erlotinib. Supplied as a solid research chemical, it is intended for preclinical pharmacology and biochemical studies to investigate EGFR signaling, drug metabolism, and related pharmacodynamic effects.
  • High purity suitable for analytical and pharmacological studies.
  • Soluble in dimethyl sulfoxide (DMSO); documented in vitro and in vivo formulations available.
  • Stable when stored as a powder at -20 °C or 4 °C for recommended storage durations.
  • Provided in small-mass quantities appropriate for research applications.
  • Synonyms and registry identifiers support cross-referencing in chemical databases.
Catalog No. 50-002-61049 Supplier Medchemexpress LLC Supplier No. HY13256A10MG
Desmethyl erlotinib is the primary active metabolite of the epidermal growth factor receptor (EGFR) inhibitor erlotinib. Supplied as a solid research chemical, it is intended for preclinical pharmacology and biochemical studies to investigate EGFR signaling, drug metabolism, and related pharmacodynamic effects.
  • High purity suitable for analytical and pharmacological studies.
  • Soluble in dimethyl sulfoxide (DMSO); documented in vitro and in vivo formulations available.
  • Stable when stored as a powder at -20 °C or 4 °C for recommended storage durations.
  • Provided in small-mass quantities appropriate for research applications.
  • Synonyms and registry identifiers support cross-referencing in chemical databases.