Learn More
Medchemexpress LLC Desmethyl ferroquine (SSR97213) | 903546-18-9 | 98.0% | 419.73 g/mol | C22H22ClFeN3 | 5 MG
SDP
Catalog No. 5000222836
Encompass_Preferred
Desmethyl ferroquine (SSR97213) is the primary active metabolite of ferroquine, an organometallic antimalarial research compound. It is supplied for laboratory use in biochemical, pharmacological, and parasitology studies where metabolite activity and compound profiling are required.
  • Primary active metabolite of ferroquine used in antimalarial research.
  • Suitable for in vitro parasite assays and metabolite profiling.
  • High purity appropriate for analytical and pharmacological studies.
  • Provided in small research quantities for laboratory experiments.
  • Includes molecular formula and molecular weight for compound characterization.
Catalog No. 50-002-22836 Supplier Medchemexpress LLC Supplier No. HY1358475MG
Desmethyl ferroquine (SSR97213) is the primary active metabolite of ferroquine, an organometallic antimalarial research compound. It is supplied for laboratory use in biochemical, pharmacological, and parasitology studies where metabolite activity and compound profiling are required.
  • Primary active metabolite of ferroquine used in antimalarial research.
  • Suitable for in vitro parasite assays and metabolite profiling.
  • High purity appropriate for analytical and pharmacological studies.
  • Provided in small research quantities for laboratory experiments.
  • Includes molecular formula and molecular weight for compound characterization.