Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Desmethyl ferroquine (SSR97213) | 903546-18-9 | 98.0% | C22H22ClFeN3 | 10MG
SDP

Catalog No. 5000258621
Encompass_Preferred

Desmethyl ferroquine (SSR97213) is the major N-monodemethylated metabolite of ferroquine, supplied as a research-grade solid for in vitro pharmacology, metabolic profiling, and analytical reference use. The material has defined purity and solubility characteristics for assay development and reference standards.

  • Primary active metabolite used in antimalarial metabolism studies.
  • High purity (98.0%).
  • Molecular formula C22H22ClFeN3; molecular weight 419.73 g/mol.
  • Soluble in DMSO (≈2.5 mg/mL); insoluble in water.
  • Available in small research-scale quantities suitable for analytical work.
  • Stable under recommended frozen storage for long-term use.

Catalog No. 50-002-58621 Supplier Medchemexpress LLC Supplier No. HY13584710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Desmethyl ferroquine (SSR97213) is the major N-monodemethylated metabolite of ferroquine, supplied as a research-grade solid for in vitro pharmacology, metabolic profiling, and analytical reference use. The material has defined purity and solubility characteristics for assay development and reference standards.

  • Primary active metabolite used in antimalarial metabolism studies.
  • High purity (98.0%).
  • Molecular formula C22H22ClFeN3; molecular weight 419.73 g/mol.
  • Soluble in DMSO (≈2.5 mg/mL); insoluble in water.
  • Available in small research-scale quantities suitable for analytical work.
  • Stable under recommended frozen storage for long-term use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.