Learn More
Medchemexpress LLC Desmethyl ketoprofen | 22071-22-3 | 99.72% | 250 MG
SDP
Catalog No. 5000341510
Encompass_Preferred
Desmethyl Ketoprofen exhibits anti-inflammatory activities and is utilized for studying angiogenesis-related disorders. It is supplied as a solid, white to off-white in appearance.
  • Has anti-inflammatory activities
  • Can be used for the study of angiogenesis-related disorders
  • Available as a solid
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, can be stored at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at ≥ 50 mg/mL
Catalog No. 50-003-41510 Supplier Medchemexpress LLC Supplier No. 250MGHY131118
Desmethyl Ketoprofen exhibits anti-inflammatory activities and is utilized for studying angiogenesis-related disorders. It is supplied as a solid, white to off-white in appearance.
  • Has anti-inflammatory activities
  • Can be used for the study of angiogenesis-related disorders
  • Available as a solid
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • In solvent, can be stored at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at ≥ 50 mg/mL