Learn More
Medchemexpress LLC Desmethyl-VS-5584 hydrochloride | 98.6% | 413.30 g/mol | C16H22Cl2N8O | 10 MG
SDP
Catalog No. 5000245238
Encompass_Preferred
Desmethyl-VS-5584 hydrochloride is the hydrochloride salt of a dimethyl analog of VS-5584, a potent and selective dual inhibitor of mTOR and PI3K with a pyrido[2,3-d]pyrimidine core. It is supplied as a high-purity research reagent for biochemical and cellular studies of PI3K/Akt/mTOR signaling and oncology-related pathways.
  • Potent and selective dual inhibitor of mTOR and PI3K.
  • Dimethyl analog with a pyrido[2,3-d]pyrimidine structure.
  • Supplied as a hydrochloride salt for research use.
  • High purity (98.6%) suitable for biochemical assays.
  • Available in multiple pack sizes for small-scale and larger studies.
Catalog No. 50-002-45238 Supplier Medchemexpress LLC Supplier No. HY101776A10MG
Desmethyl-VS-5584 hydrochloride is the hydrochloride salt of a dimethyl analog of VS-5584, a potent and selective dual inhibitor of mTOR and PI3K with a pyrido[2,3-d]pyrimidine core. It is supplied as a high-purity research reagent for biochemical and cellular studies of PI3K/Akt/mTOR signaling and oncology-related pathways.
  • Potent and selective dual inhibitor of mTOR and PI3K.
  • Dimethyl analog with a pyrido[2,3-d]pyrimidine structure.
  • Supplied as a hydrochloride salt for research use.
  • High purity (98.6%) suitable for biochemical assays.
  • Available in multiple pack sizes for small-scale and larger studies.