Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Desmethyl-VS-5584 hydrochloride | 98.6% | 413.30 g/mol | C16H22Cl2N8O | 25 MG
SDP

Catalog No. 5000249053
Encompass_Preferred

Desmethyl-VS-5584 hydrochloride is a dimethyl analog of VS-5584 that functions as a potent and selective dual inhibitor of mTOR and PI3K, intended for research use. It contains a pyrido[2,3-d]pyrimidine core and is supplied as a light yellow solid with high reported purity.

  • Potent, selective mTOR and PI3K dual inhibitor.
  • High purity (98.6%) suitable for research applications.
  • Molecular weight 413.30 g/mol and formula C16H22Cl2N8O.
  • Solid, light yellow to yellow appearance.
  • Available in small lab-scale quantities (e.g., 25 mg).
  • Storage: store sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-49053 Supplier Medchemexpress LLC Supplier No. HY101776A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Desmethyl-VS-5584 hydrochloride is a dimethyl analog of VS-5584 that functions as a potent and selective dual inhibitor of mTOR and PI3K, intended for research use. It contains a pyrido[2,3-d]pyrimidine core and is supplied as a light yellow solid with high reported purity.

  • Potent, selective mTOR and PI3K dual inhibitor.
  • High purity (98.6%) suitable for research applications.
  • Molecular weight 413.30 g/mol and formula C16H22Cl2N8O.
  • Solid, light yellow to yellow appearance.
  • Available in small lab-scale quantities (e.g., 25 mg).
  • Storage: store sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.