Learn More
Cayman Chemical Desmethylcitalopram-d3 hydro
SDP
Catalog No. 502972173
Encompass_Preferred
An internal standard for the quantification of desmethylcitalopram by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-297-2173 Supplier Cayman Chemical Supplier No. 15840500 UG
An internal standard for the quantification of desmethylcitalopram by GC- or LC-MS; intended for research and forensic applications