Learn More
Cayman Chemical Desmethylene Paroxetine (hydrochloride), Purity: ≥98%, CAS: 1394861-12-1, 50 mg
SDP
Catalog No. 502971039
Encompass_Preferred
A major urinary metabolite of the selective serotonin reuptake inhibitor paroxetine; may be used as a control in LC/MS or GC/MS paroxetine testing applications
Catalog No. 50-297-1039 Supplier Cayman Chemical Supplier No. 1590850 MG
A major urinary metabolite of the selective serotonin reuptake inhibitor paroxetine; may be used as a control in LC/MS or GC/MS paroxetine testing applications