Learn More
OriGene Desmin (DES) CytoSection, Positive Control For IHC, ICC, & ISH, 25 Slides
SDP
Catalog No. NC3312162
Encompass_Preferred
The Origene Technologies Desmin (DES) CytoSection is a crucial tool for researchers, providing validated positive controls for detecting Desmin expression in immunohistochemical, immunocytochemical, and *in situ* hybridization applications. This product includes 25 FFPE slides containing HEK293T cells with transient overexpression of human Desmin, ensuring reliable and accurate experimental results, distinguishing it from other controls with its specific targeted application.
Catalog No. NC3312162 Supplier OriGene Supplier No. TS405685P5
The Origene Technologies Desmin (DES) CytoSection is a crucial tool for researchers, providing validated positive controls for detecting Desmin expression in immunohistochemical, immunocytochemical, and *in situ* hybridization applications. This product includes 25 FFPE slides containing HEK293T cells with transient overexpression of human Desmin, ensuring reliable and accurate experimental results, distinguishing it from other controls with its specific targeted application.