Learn More
Bioworld DESMOPRESSIN
Supplier:  Bioworld 220600541
Encompass_Preferred
Desmopressin Description : Desmopressin also called ADH (Anti-Diuretic Hormone) has a molecular formula of C46H64N14O12S2 , Mpr-Tyr-Phe-Gln-Asn-Cys-Pro-D-Arg-Gly-NH2 having a Mw of 1069.23 is the first vasopressin analog with a very hi...
Catalog No. 50-253-151