Learn More
TARGETMOL CHEMICALS INC DESMOPRESSIN ACETATE 166 10MG
SDP
Catalog No. 502579805
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Desmopressin acetate (16679-58-6 free base) (DDAVP) is the synthetic analog of the antidiuretic hormone arginine vasopressin. Vasopressin Receptor The antidiuretic properties of desmopressin have led to its use in polyuric conditions including primary nocturnal enuresis nocturia and diabetes insipidus. Desmopressin acetate (16679-58-6 free base) works by limiting the amount of water that is eliminated in the urine. Desmopressin acetate (16679-58-6 free base) binds to V2 receptors in renal collecting ducts increasing water reabsorption. It also stimulates the release of von Willebrand factor from endothelial cells by acting on the V2 receptor. purity: 100%
Catalog No. 50-257-9805 Supplier TARGETMOL CHEMICALS INC Supplier No. T514410MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Desmopressin acetate (16679-58-6 free base) (DDAVP) is the synthetic analog of the antidiuretic hormone arginine vasopressin. Vasopressin Receptor The antidiuretic properties of desmopressin have led to its use in polyuric conditions including primary nocturnal enuresis nocturia and diabetes insipidus. Desmopressin acetate (16679-58-6 free base) works by limiting the amount of water that is eliminated in the urine. Desmopressin acetate (16679-58-6 free base) binds to V2 receptors in renal collecting ducts increasing water reabsorption. It also stimulates the release of von Willebrand factor from endothelial cells by acting on the V2 receptor. purity: 100%