Learn More
Selleck Chemical LLC Desmopressin Acetate-5mg
SDP
Supplier:  Selleck Chemical LLC P10845MG
Encompass_Preferred
Desmopressin is a synthetic octapeptide, and an analogue of human hormone arginine vasopressin with antidiuretic and coagulant activities. It is a selective agonist for the vasopressin V2 receptor (V2R).
Catalog No. 50-238-9619