Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Desmorpholinyl navitoclax-NH-Me | 2365172-82-1 | MFCD32857149 | 98.6% | 918.55 | C44H51ClF3N5O5S3 | 5 MG
SDP

Catalog No. 5000262235
Encompass_Preferred

Desmorpholinyl Navitoclax-NH-Me is a navitoclax-derived BCL-XL inhibitor provided as a purified research chemical and synthetic building block for medicinal chemistry and PROTAC development. It is supplied in multiple small-scale pack sizes for laboratory use and includes characterization data for research applications.

  • High purity suitable for biochemical and medicinal research (reported 98.64%).
  • Available in multiple small pack sizes for screening and synthesis work.
  • Molecular weight 918.55 and defined molecular formula for accurate reagent selection.
  • SMILES and structural data provided to support cheminformatics workflows.
  • Applicable as a building block in PROTAC synthesis and BCL-XL targeted studies.

Catalog No. 50-002-62235 Supplier Medchemexpress LLC Supplier No. HY1312325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Desmorpholinyl Navitoclax-NH-Me is a navitoclax-derived BCL-XL inhibitor provided as a purified research chemical and synthetic building block for medicinal chemistry and PROTAC development. It is supplied in multiple small-scale pack sizes for laboratory use and includes characterization data for research applications.

  • High purity suitable for biochemical and medicinal research (reported 98.64%).
  • Available in multiple small pack sizes for screening and synthesis work.
  • Molecular weight 918.55 and defined molecular formula for accurate reagent selection.
  • SMILES and structural data provided to support cheminformatics workflows.
  • Applicable as a building block in PROTAC synthesis and BCL-XL targeted studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.