Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmosine, 99.5%, MP Biomedicals™

Catalog No. ICN19137801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19137801 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19137801 Supplier MP Biomedicals, Inc Supplier No. 0219137801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity desmosine derived from bovine neck ligament, used as a biomarker for elastin degradation and cross-linking studies.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11003-57-9
Molecular Formula C24H40N5O8
Molecular Weight (g/mol) 526.61
InChI Key VEVRNHHLCPGNDU-UHFFFAOYNA-O
Synonym desmosine, norleucine, 6-4-4-amino-4-carboxybutyl-3,5-bis 3-amino-3-carboxypropyl pyridinio, 2-amino-6-3,5-bis 3-amino-4-hydroxy-4-oxobutyl-4-4-amino-5-hydroxy-5-oxopentyl pyridin-1-ium-1-yl hexanoic acid
PubChem CID 15942890
ChEBI CHEBI:37628

Specifications

Color Yellow
Assay Percent Range 0.995
Quantity 1 mg
Formula Weight 526
Percent Purity 99%
Grade Not Available
Physical Form Solid
Chemical Name or Material Desmosine
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.