Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmosine, MP Biomedicals™

Catalog No. mp219137805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219137805 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219137805 Supplier MP Biomedicals, Inc Supplier No. 0219137805
Only null left
Add to Cart
Add to Cart

Responsible for the cross-linking of elastin, leading to the formation of insoluble fibers. Desmosine, MP Biomedicals is a tetrafunctional, pyridinium ring-containing amino acids involved in elastin cross-linking. Urinary excretion of desmosine is an indi

A desmosine cross-link is formed from three allysyl side chains plus one unaltered lysyl side chain from the same or neighbouring polypeptides. Desmosine is responsible for the cross-linking of elastin. Urinary excretion of desmosine is a useful indicator of elastin breakdown resulting from increased elastase activity. It also acts as a biomarker of elastin degradation in chronic obstructive pulmonary disease.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11003-57-9
Molecular Formula C24H40N5O8+
Molecular Weight (g/mol) 526.611
InChI Key VEVRNHHLCPGNDU-MUGJNUQGSA-O
Synonym unii-4mol2j0tly, desmosine, 4mol2j0tly, +-desmosine, 4-4s-4-amino-4-carboxybutyl-1-5s-5-amino-5-carboxypentyl-3,5-bis 3s-3-amino-3-carboxypropyl pyridinium, pyridinium, 4-4-amino-4-carboxybutyl-1-5-amino-5-carboxypentyl-3,5-bis 3-amino-3-carboxypropyl, pyridinium, 4-4-amino-4-carboxybutyl-1-5-amino-5-carboxypentyl-3,5-bis 3-amino-3-carboxypropyl-van
PubChem CID 15942890
ChEBI CHEBI:37628
IUPAC Name (2S)-2-amino-6-[4-[(4S)-4-amino-4-carboxybutyl]-3,5-bis[(3S)-3-amino-3-carboxypropyl]pyridin-1-ium-1-yl]hexanoic acid
SMILES C1=C(C(=C(C=[N+]1CCCCC(C(=O)O)N)CCC(C(=O)O)N)CCCC(C(=O)O)N)CCC(C(=O)O)N

Specifications

Quantity 5 mg
Formula Weight 526 (anhydrous)
Percent Purity 99%
Physical Form Solid
Recommended Storage 4°C, desiccate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.