Learn More
Medchemexpress LLC Desmosterol | 313-04-2 | MFCD00056432 | 99.8% | 384.64 g/mol | C27H44O | 5 MG
SDP
Catalog No. 5000263440
Encompass_Preferred
Desmosterol is an endogenous sterol and direct precursor to cholesterol in the Bloch biosynthetic pathway. It is used as a research reagent to probe cholesterol biosynthesis, nuclear receptor signaling, and lipid metabolism in biochemical and cell-based studies.
  • High purity (>99.8%) suitable for analytical and biological assays.
  • Molecular weight 384.64 g/mol; formula C27H44O.
  • Acts as an LXR activator and SREBP pathway probe.
  • Provided as a 5 mg reagent for small-scale experiments.
  • Compatible with biochemical assays and lipid metabolism studies.
Catalog No. 50-002-63440 Supplier Medchemexpress LLC Supplier No. HY1132245MG
Desmosterol is an endogenous sterol and direct precursor to cholesterol in the Bloch biosynthetic pathway. It is used as a research reagent to probe cholesterol biosynthesis, nuclear receptor signaling, and lipid metabolism in biochemical and cell-based studies.
  • High purity (>99.8%) suitable for analytical and biological assays.
  • Molecular weight 384.64 g/mol; formula C27H44O.
  • Acts as an LXR activator and SREBP pathway probe.
  • Provided as a 5 mg reagent for small-scale experiments.
  • Compatible with biochemical assays and lipid metabolism studies.