Learn More
Kyfora Bio Desmosterol 5 MG
Catalog No. 502559795
Encompass_Preferred
Desmosterol (24-dehydro cholesterol) is a sterol lipid with a backbone similar to cholesterol. It is the precursor to cholesterol in the Bloch biosynthesis pathway. It is known to be a regulator of lipid metabolism. Desmosterol can be used in the study of cellular membranes and lipid metabolism.Key application(s): Lipid metabolism, cellular membranes
Catalog No. 50-255-9795 Supplier Kyfora Bio Supplier No. 145225
Desmosterol (24-dehydro cholesterol) is a sterol lipid with a backbone similar to cholesterol. It is the precursor to cholesterol in the Bloch biosynthesis pathway. It is known to be a regulator of lipid metabolism. Desmosterol can be used in the study of cellular membranes and lipid metabolism.Key application(s): Lipid metabolism, cellular membranes