Learn More
Cayman Chemical Desogestrel 25mg
SDP
Catalog No. 502964386
Encompass_Preferred
A synthetic progestogen; inhibits ovulation and prevents fertilization in rabbits and mice; inhibits rhodamine123 efflux, a measure of P-glycoprotein activity, ex vivo in human lymphocytes and CD8+ T cells in a dose-dependent manner
Catalog No. 50-296-4386 Supplier Cayman Chemical Supplier No. 2365125 MG
A synthetic progestogen; inhibits ovulation and prevents fertilization in rabbits and mice; inhibits rhodamine123 efflux, a measure of P-glycoprotein activity, ex vivo in human lymphocytes and CD8+ T cells in a dose-dependent manner