Learn More
Medchemexpress LLC Desonide-13C3 | >98.0% | 419.49 | C21 13C3 H32 O6 | 1 MG
SDP
Catalog No. 5000284384
Encompass_Preferred
Desonide-13C3 is the 13C-labeled isotopologue of desonide provided for research use. It is intended as a tracer, internal standard, or analytical reference in studies of glucocorticoid receptor activity, inflammatory disease models, and Huntington's disease research. Supplied as a solid in small vials and accompanied by a certificate of analysis with storage recommendations.
  • 13C-labeled isotopologue suitable for tracer and mass spectrometry internal standard use.
  • High isotopic purity supports accurate quantitation in analytical workflows.
  • Available in small-scale quantities for method development and validation.
  • Supplied as a solid; follow the certificate of analysis for storage and handling.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-84384 Supplier Medchemexpress LLC Supplier No. HYW6539891MG
Desonide-13C3 is the 13C-labeled isotopologue of desonide provided for research use. It is intended as a tracer, internal standard, or analytical reference in studies of glucocorticoid receptor activity, inflammatory disease models, and Huntington's disease research. Supplied as a solid in small vials and accompanied by a certificate of analysis with storage recommendations.
  • 13C-labeled isotopologue suitable for tracer and mass spectrometry internal standard use.
  • High isotopic purity supports accurate quantitation in analytical workflows.
  • Available in small-scale quantities for method development and validation.
  • Supplied as a solid; follow the certificate of analysis for storage and handling.
  • Intended for research use only; not for human or clinical use.