Learn More
Cayman Chemical Desotamide, Purity: ≥95%, CAS: 194660-14-5, 2.5 mg
SDP
Catalog No. 502941141
Encompass_Preferred
A cyclic hexapeptide antibiotic; active against S. aureus, S. pneumoniae, and MRSE (MICs = 16, 12.5, and 32 μg/ml, respectively)
Catalog No. 50-294-1141 Supplier Cayman Chemical Supplier No. 281292.5 MG
A cyclic hexapeptide antibiotic; active against S. aureus, S. pneumoniae, and MRSE (MICs = 16, 12.5, and 32 μg/ml, respectively)