Learn More
Cayman Chemical Desoximetasone, Purity: ≥98%, CAS: 382-67-2, 10 mg
SDP
Catalog No. 502995038
Encompass_Preferred
A glucocorticoid; reverses TPA-induced increases in epidermal cell proliferation in SENCAR mice (0.001-0.1 mg topically); reduces TPA-induced expression of IL-6, COX-2, iNOS, and c-Jun mRNA levels in the skin of SENCAR mice
Catalog No. 50-299-5038 Supplier Cayman Chemical Supplier No. 2394510 MG
A glucocorticoid; reverses TPA-induced increases in epidermal cell proliferation in SENCAR mice (0.001-0.1 mg topically); reduces TPA-induced expression of IL-6, COX-2, iNOS, and c-Jun mRNA levels in the skin of SENCAR mice