Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Desoximetasone | 382-67-2 | MFCD00083301 | 99.0% | 376.46 | C22H29FO4 | 10 MG
SDP

Catalog No. 5000298405
Encompass_Preferred

Desoximetasone analytical standard is a characterized reference material of the corticosteroid desoximetasone intended for research and analytical applications, including assay development, method validation, and quality control. The material is supplied as a solid with defined composition and documented analytical data to support accurate identification and quantification in laboratory workflows.

  • Intended use: analytical standard for research and method validation.
  • Purity: 98.97% as reported by the supplier.
  • Molecular weight: 376.46 g/mol, for mass-based calculations.
  • Chemical formula: C22H29FO4, confirming compound identity.
  • Physical form: solid for stable handling and storage.
  • Storage: follow the conditions specified in the certificate of analysis.

Catalog No. 50-002-98405 Supplier Medchemexpress LLC Supplier No. HY17570R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Desoximetasone analytical standard is a characterized reference material of the corticosteroid desoximetasone intended for research and analytical applications, including assay development, method validation, and quality control. The material is supplied as a solid with defined composition and documented analytical data to support accurate identification and quantification in laboratory workflows.

  • Intended use: analytical standard for research and method validation.
  • Purity: 98.97% as reported by the supplier.
  • Molecular weight: 376.46 g/mol, for mass-based calculations.
  • Chemical formula: C22H29FO4, confirming compound identity.
  • Physical form: solid for stable handling and storage.
  • Storage: follow the conditions specified in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.