Learn More
Medchemexpress LLC Desoximetasone (Standard) | 382-67-2 | 98.97% | 376.46 | 100 MG
SDP
Catalog No. 5000386917
Encompass_Preferred
Desoximetasone (Standard) is the analytical standard of Desoximetasone. This product is intended for research and analytical applications. Desoximetasone is a corticosteroid and a substrate of cytochrome P450 3A4. Desoximetasone has anti-inflammatory, anti-allergic, and immunosuppressive effects. Desoximetasone can be used in the research of various skin diseases, such as skin allergies, atopic dermatitis, and psoriasis.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-86917 Supplier Medchemexpress LLC Supplier No. HY17570R100MG
Desoximetasone (Standard) is the analytical standard of Desoximetasone. This product is intended for research and analytical applications. Desoximetasone is a corticosteroid and a substrate of cytochrome P450 3A4. Desoximetasone has anti-inflammatory, anti-allergic, and immunosuppressive effects. Desoximetasone can be used in the research of various skin diseases, such as skin allergies, atopic dermatitis, and psoriasis.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.