Learn More
Medchemexpress LLC Desoximetasone (Standard) | 382-67-2 | 98.97% | 376.46 | 5 MG
SDP
Catalog No. 5000387157
Encompass_Preferred
Desoximetasone (Standard) is the analytical standard of Desoximetasone. This product is intended for research and analytical applications. It is a corticosteroid and a substrate of cytochrome P450 3A4, with anti-inflammatory, anti-allergic, and immunosuppressive effects. It can be used in the research of various skin diseases, such as skin allergies, atopic dermatitis, and psoriasis.
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-87157 Supplier Medchemexpress LLC Supplier No. HY17570R5MG
Desoximetasone (Standard) is the analytical standard of Desoximetasone. This product is intended for research and analytical applications. It is a corticosteroid and a substrate of cytochrome P450 3A4, with anti-inflammatory, anti-allergic, and immunosuppressive effects. It can be used in the research of various skin diseases, such as skin allergies, atopic dermatitis, and psoriasis.
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.