Learn More
Medchemexpress LLC Desoximetasone (Standard) | 382-67-2 | 99.0% | 376.46 | 25 MG
SDP
Catalog No. 5000384464
Encompass_Preferred
Desoximetasone (Standard) is the analytical standard of Desoximetasone and is intended for research and analytical applications. It is a corticosteroid and a substrate of cytochrome P450 3A4, possessing anti-inflammatory, anti-allergic, and immunosuppressive effects. It is utilized in the research of various skin diseases, including skin allergies, atopic dermatitis, and psoriasis.
  • Analytical standard for Desoximetasone.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Possesses anti-inflammatory, anti-allergic, and immunosuppressive effects.
  • Applicable for research in skin diseases such as skin allergies, atopic dermatitis, and psoriasis.
Catalog No. 50-003-84464 Supplier Medchemexpress LLC Supplier No. 25MGHY17570R
Desoximetasone (Standard) is the analytical standard of Desoximetasone and is intended for research and analytical applications. It is a corticosteroid and a substrate of cytochrome P450 3A4, possessing anti-inflammatory, anti-allergic, and immunosuppressive effects. It is utilized in the research of various skin diseases, including skin allergies, atopic dermatitis, and psoriasis.
  • Analytical standard for Desoximetasone.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Possesses anti-inflammatory, anti-allergic, and immunosuppressive effects.
  • Applicable for research in skin diseases such as skin allergies, atopic dermatitis, and psoriasis.