Learn More
Medchemexpress LLC Desoxo-narchinol A | 53859-06-6 | 1 MG
SDP
Catalog No. 5000419135
Encompass_Preferred
Desoxo-narchinol A is an orally active and potent anti-inflammatory agent, isolated from the roots and rhizomes of Nardostachys jatamansi. It is used for septic shock and inflammatory diseases research, effectively inhibiting tissue injury and the production of pro-inflammatory cytokines such as IL-1β, IL-6, and TNF-α.
  • Orally active and potent anti-inflammatory agent
  • Isolated from the roots and rhizomes of Nardostachys jatamansi
  • Research applications include septic shock and inflammatory diseases
  • Inhibits the production of key pro-inflammatory cytokines and mediators (IL-1β, IL-6, TNF-α, PGE2, nitric oxide)
  • Demonstrates efficacy in reducing mortality in endotoxin shock models
  • Exhibits oral bioavailability in animal models
Catalog No. 50-004-19135 Supplier Medchemexpress LLC Supplier No. HYN84351MG
Desoxo-narchinol A is an orally active and potent anti-inflammatory agent, isolated from the roots and rhizomes of Nardostachys jatamansi. It is used for septic shock and inflammatory diseases research, effectively inhibiting tissue injury and the production of pro-inflammatory cytokines such as IL-1β, IL-6, and TNF-α.
  • Orally active and potent anti-inflammatory agent
  • Isolated from the roots and rhizomes of Nardostachys jatamansi
  • Research applications include septic shock and inflammatory diseases
  • Inhibits the production of key pro-inflammatory cytokines and mediators (IL-1β, IL-6, TNF-α, PGE2, nitric oxide)
  • Demonstrates efficacy in reducing mortality in endotoxin shock models
  • Exhibits oral bioavailability in animal models