Learn More
Medchemexpress LLC Desoxyanisoin 100g
SDP
Catalog No. 5000205418
Encompass_Preferred
Desoxyanisoin is a precursor of 4 4-bishydroxydeoxybenzoin (BHDB) Desoxyanisoin can be used in the study of epoxy thermoset reinforcement materials[1]
Catalog No. 50-002-05418 Supplier Medchemexpress LLC Supplier No. HYW097754100G
Desoxyanisoin is a precursor of 4 4-bishydroxydeoxybenzoin (BHDB) Desoxyanisoin can be used in the study of epoxy thermoset reinforcement materials[1]