Learn More
Medchemexpress LLC Desoxycarbadox-d3 | 1448350-02-4 | >95.0% | 233.24 | C11H7D3N4O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY136428S5MG
Encompass_Preferred
Desoxycarbadox-d3 is the deuterium-labeled analogue of desoxycarbadox provided as a small-quantity analytical reference standard for research. It is intended as an internal standard or reference material for trace analysis, method development, and quantitative mass spectrometry workflows involving desoxycarbadox and related metabolites. Supplied as a 5 mg vial; product is for research use only.
  • Deuterium-labeled internal standard for analytical assays.
  • Intended for quantitative mass spectrometry and method development.
  • Supplied as a 5 mg vial for small-scale analyses.
  • Purity >95.0% (HPLC) as reported by reference suppliers.
  • For research use only; not for diagnostic or therapeutic use.
Catalog No. 50-002-22900