Learn More
Medchemexpress LLC Desoxycorticosterone pivalate | 808-48-0 | MFCD00199992 | 99.6% | 414.58 g/mol | C26H38O4 | 25 MG
SDP
Catalog No. 5000242133
Encompass_Preferred
Desoxycorticosterone pivalate (DOCP) is a mineralocorticoid hormone analog used in pharmacological and biochemical research, commonly in studies of mineralocorticoid receptor activity and in canine hypoadrenocorticism models. It is supplied as a research-grade powder with established analytical characterization and typical solvent compatibility for laboratory formulations.
  • High purity suitable for analytical and research use.
  • Supplied as a powder for flexible formulation and dosing.
  • Compatible with DMSO for preparation of stock solutions.
  • Storage stability documented for powder and solution states.
  • Analytical support available, including COA, H NMR, and LC-MS data.
Catalog No. 50-002-42133 Supplier Medchemexpress LLC Supplier No. HY10791725MG
Desoxycorticosterone pivalate (DOCP) is a mineralocorticoid hormone analog used in pharmacological and biochemical research, commonly in studies of mineralocorticoid receptor activity and in canine hypoadrenocorticism models. It is supplied as a research-grade powder with established analytical characterization and typical solvent compatibility for laboratory formulations.
  • High purity suitable for analytical and research use.
  • Supplied as a powder for flexible formulation and dosing.
  • Compatible with DMSO for preparation of stock solutions.
  • Storage stability documented for powder and solution states.
  • Analytical support available, including COA, H NMR, and LC-MS data.